Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019005.3 | NP_061878.3 | 4 | 1213 | ||
XM_005249780.3 | XP_005249837.1 | 3 | 1241 | |||
XM_005249781.3 | XP_005249838.1 | 2 | 1057 | |||
XM_005249782.3 | XP_005249839.1 | 2 | 1442 | |||
XM_005249784.3 | XP_005249841.1 | 3 | 1241 | |||
XM_011515432.2 | XP_011513734.1 | 2 | 1039 | |||
XM_017012361.1 | XP_016867850.1 | 3 | 1408 | |||
XM_017012362.1 | XP_016867851.1 | 2 | 1057 | |||
GenBank mRNA | AK000330.1 | - | 3 | 1086 | ||
AK294561.1 | - | 4 | 1172 | |||
BC140833.1 | - | 3 | 1063 | 9021701 | ||
BC140834.1 | - | 2 | 965 | 9021702 |