Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LRCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243963.1 | NP_001230892.1 | 5 | 777 | ||
NM_020871.3 | NP_065922.3 | 5 | 777 | |||
XM_006724724.3 | XP_006724787.2 | 5 | 780 | |||
XM_017029696.1 | XP_016885185.1 | 5 | 786 | |||
XM_017029697.1 | XP_016885186.1 | 5 | 787 | |||
GenBank mRNA | BC125224.1 | - | 5 | 770 | 40124466 | |
BX647193.1 | - | 5 | 777 |