Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020961.3 | NP_066012.1 | 11 | 1536 | ||
XM_005263166.4 | XP_005263223.1 | 10 | 1320 | |||
XM_011532162.2 | XP_011530464.1 | 11 | 1370 | |||
GenBank mRNA | AB046847.1 | - | 11 | 1491 | ||
AK055555.1 | - | 11 | 1514 | |||
AK296056.1 | - | 9 | 1156 | |||
BC006565.2 | - | 11 | 1487 | 3010654 | ||
BC007449.2 | - | 11 | 1487 | 3010654 |