Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERPUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022373.4 | NP_071768.3 | 7 | 1453 | ||
XM_006715765.1 | XP_006715828.1 | 5 | 1261 | |||
XM_011515473.1 | XP_011513775.1 | 4 | 1459 | |||
XM_017012506.1 | XP_016867995.1 | 6 | 1651 | |||
GenBank mRNA | AK025966.1 | - | 2 | 309 | ||
AK055708.1 | - | 6 | 1600 | |||
AK315685.1 | - | 6 | 1077 | |||
BC005091.1 | - | 7 | 1259 | 3840918 | ||
BC020264.1 | - | 7 | 1410 | 4878523 |