Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022483.4 | NP_071928.2 | 3 | 1380 | ||
XM_005248333.4 | XP_005248390.1 | 4 | 2248 | |||
XM_005248335.3 | XP_005248392.1 | 4 | 1532 | |||
XM_006714486.3 | XP_006714549.1 | 3 | 1852 | |||
XM_011514074.2 | XP_011512376.1 | 5 | 2624 | |||
XM_011514075.2 | XP_011512377.1 | 5 | 2620 | |||
GenBank mRNA | AK025310.1 | - | 3 | 1349 | ||
AL137306.1 | - | 1 | 635 | |||
BC013351.2 | - | 4 | 1569 | 3621124 | ||
BC063832.1 | - | 2 | 1326 | 6178186 | ||
BC073852.1 | - | 2 | 1313 |