Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024332.3 | NP_077308.1 | 8, 9 | 739 | ||
XM_005274751.4 | XP_005274808.1 | 8, 9 | 752 | |||
GenBank mRNA | AK298886.1 | - | 7, 8 | 574 | ||
AK313544.1 | - | 8, 9 | 634 | |||
AY438030.1 | - | 8, 9 | 631 | |||
BC002999.1 | - | 8, 9 | 633 | 2821917 | ||
BC006540.2 | - | 8, 9 | 633 | 2821917 | ||
S68015.1 | - | 8, 9 | 664 |