Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024593.3 | NP_078869.1 | 6 | 2014 | ||
NR_024605.1 | 6 | 1730 | ||||
XM_005251303.1 | XP_005251360.1 | 7 | 1890 | |||
XM_011517590.2 | XP_011515892.1 | 7 | 2144 | |||
XM_011517591.2 | XP_011515893.1 | 7 | 2117 | |||
XM_017013826.1 | XP_016869315.1 | 6 | 1791 | |||
XM_017013827.1 | XP_016869316.1 | 8 | 2267 | |||
GenBank mRNA | AK021829.1 | - | 7 | 1761 | ||
BC025676.1 | - | 6 | 1935 | 5203436 | ||
DB536362.1 | - | 1 | 333 | |||
DB552638.1 | - | 1 | 342 |