Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178055.1 | NP_001171526.1 | 13 | 1256 | ||
NM_001178056.1 | NP_001171527.1 | 12 | 1232 | |||
NM_024615.3 | NP_078891.2 | 12 | 1232 | |||
XM_005248596.4 | XP_005248653.1 | 13 | 1612 | |||
XM_011543631.2 | XP_011541933.1 | 13 | 1145 | |||
XM_011543632.2 | XP_011541934.2 | 13 | 1192 | |||
XM_011543633.2 | XP_011541935.2 | 13 | 1192 | |||
XM_011543634.2 | XP_011541936.1 | 14 | 1741 | |||
XM_011543635.2 | XP_011541937.1 | 14 | 1262 | |||
XM_011543636.2 | XP_011541938.1 | 13 | 1126 | |||
XM_011543640.2 | XP_011541942.1 | 10 | 1016 | |||
XM_011543641.2 | XP_011541943.1 | 11 | 993 | |||
XM_011543642.2 | XP_011541944.1 | 9 | 980 | |||
XM_011543643.2 | XP_011541945.1 | 10 | 1421 | |||
XM_017009850.1 | XP_016865339.1 | 12 | 1692 | |||
XM_017009851.1 | XP_016865340.1 | 11 | 1132 | |||
GenBank mRNA | AK024961.1 | - | 2 | 302 | ||
AK225188.1 | - | 2 | 302 | |||
AK289369.1 | - | 12 | 1215 | |||
AK298680.1 | - | 11 | 1095 | |||
AK314466.1 | - | 13 | 1256 | |||
AL834477.1 | - | 12 | 1191 | |||
BC037386.1 | - | 10 | 1090 | 4830820 | ||
BC075801.1 | - | 12 | 1191 | 5226828 |