Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002814.2 | NP_001002814.2 | 6 | 3783 | ||
NM_025151.4 | NP_079427.4 | 5 | 1881 | |||
GenBank mRNA | AF368294.1 | - | 5 | 1851 | ||
AK026177.1 | - | 2 | 122 | |||
AK092296.1 | - | 4 | 1189 | |||
AK122583.1 | - | 3 | 1922 | |||
AK124422.1 | - | 1 | 681 | |||
AK226111.1 | - | 6 | 3473 | |||
AK291781.1 | - | 5 | 1878 | |||
AL832155.1 | - | 3 | 1364 | |||
AL833169.1 | - | 3 | 1192 | |||
AY280966.1 | - | 3 | 2204 | |||
AY280968.1 | - | 6 | 3740 | |||
AY280969.1 | - | 2 | 2953 | |||
BC077720.1 | - | 5 | 1851 | 6537110 | ||
DQ083995.1 | - | 5 | 1605 | |||
DQ236342.1 | - | 5 | 1851 |