Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025159.2 | NP_079435.1 | 3 | 941 | ||
GenBank mRNA | AK021639.1 | - | 3 | 822 | ||
BC020611.1 | - | 3 | 869 | 4768735 | ||
BX281759.1 | - | 1 | 401 |