Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170649.1 | NP_001164120.1 | 12 | 2063 | ||
NM_001170650.1 | NP_001164121.1 | 11 | 1937 | |||
NM_031206.4 | NP_112483.1 | 13 | 2114 | |||
XM_005262301.1 | XP_005262358.1 | 13 | 2096 | |||
XM_011531045.1 | XP_011529347.1 | 12 | 1963 | |||
XM_017029877.1 | XP_016885366.1 | 12 | 2045 | |||
XM_017029879.1 | XP_016885368.1 | 12 | 1989 | |||
XM_017029880.1 | XP_016885369.1 | 11 | 1938 | |||
GenBank mRNA | AF116730.1 | - | 13 | 2052 | ||
AK022587.1 | - | 13 | 2064 | |||
AK054817.1 | - | 3 | 1730 | |||
AK074087.1 | - | 12 | 4829 | |||
AK097383.1 | - | 11 | 1887 | |||
AK125760.1 | - | 2 | 1591 | |||
AK225059.1 | - | 12 | 1996 | |||
BC014545.1 | - | 13 | 2098 | 4079376 | ||
BC018610.1 | - | 12 | 2011 | 3140243 | ||
BC019302.1 | - | 12 | 2008 | 2822893 |