Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160372.3 | NP_001153844.1 | 13, 14 | 2109 | ||
NM_001321646.1 | NP_001308575.1 | 12, 13 | 2082 | |||
NM_031466.7 | NP_113654.4 | 13, 14 | 2284 | |||
XM_011517326.2 | XP_011515628.1 | 12, 13 | 2257 | |||
XM_011517328.2 | XP_011515630.1 | 13, 14 | 2284 | |||
XM_011517330.2 | XP_011515632.1 | 5, 6 | 649 | |||
XM_017013893.1 | XP_016869382.1 | 13, 14 | 2284 | |||
XM_017013894.1 | XP_016869383.1 | 6, 7 | 985 | |||
GenBank mRNA | AB067469.1 | - | 13, 14 | 2059 | ||
AK026394.1 | - | 13, 14 | 2101 | |||
AK027689.1 | - | 2, 3 | 209 | |||
AK128755.1 | - | 13, 14 | 2285 | |||
AK308815.1 | - | 2, 3 | 380 | |||
AL833973.1 | - | 11, 12 | 1384 | |||
AY190606.1 | - | 11, 12 | 1363 | |||
BC006206.2 | - | 11, 12 | 1505 | 3538806 |