Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134340.1 | NP_001127812.1 | 3 | 634 | ||
NM_032302.3 | NP_115678.1 | 2 | 1243 | |||
GenBank mRNA | BC004308.1 | - | 2 | 822 | 3627097 | |
BC027171.1 | - | 2 | 1244 | 4344108 | ||
CN261807.1 | - | 2 | 628 |