Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032882.5 | NP_116271.3 | 2 | 1399 | ||
GenBank mRNA | AK055322.1 | - | 2 | 1331 | ||
AK289879.1 | - | 2 | 1377 | |||
BC007631.1 | - | 1 | 1131 | 3507248 |