Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033339.1 | 2 | 1740 | |||
GenBank mRNA | AL390154.1 | - | 1 | 791 | ||
BC013775.1 | - | 1 | 843 | |||
BC111466.1 | - | 1 | 1188 |