Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130899.2 | NP_570969.2 | 2 | 998 | ||
GenBank mRNA | AK057543.1 | - | 2 | 994 | ||
AY973323.1 | - | 2 | 898 | |||
BC022035.1 | - | 2 | 1006 | 4826350 | ||
BC025397.1 | - | 2 | 999 | 4827541 | ||
BC025409.1 | - | 2 | 999 | 4825198 | ||
BC025998.1 | - | 2 | 999 | 4836822 |