Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287580.1 | NP_001274509.1 | 3, 4 | 1400 | ||
NM_022128.2 | NP_071411.1 | 2, 3 | 281 | |||
GenBank mRNA | AJ404857.1 | - | 2, 3 | 241 | ||
AK300989.1 | - | 2, 3 | 235 | |||
AY643715.1 | - | 2, 3 | 273 | |||
BC017425.1 | - | 2, 3 | 283 | 4694872 | ||
BX648909.1 | - | 3, 4 | 1371 |