Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM91A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317917.1 | NP_001304846.1 | 14 | 1535 | ||
NM_001317918.1 | NP_001304847.1 | 14 | 1528 | |||
NM_144963.3 | NP_659400.2 | 14 | 1528 | |||
GenBank mRNA | AK074370.1 | - | 9 | 835 | ||
AK300603.1 | - | 14 | 1511 | |||
AL832830.1 | - | 3 | 259 | |||
BC144630.1 | - | 14 | 1328 | |||
DQ228141.1 | - | 14 | 1325 |