Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031745.4 | NP_001026915.1 | 5 | 567 | ||
NM_144968.3 | NP_659405.1 | 5 | 567 | |||
XM_005261988.3 | XP_005262045.1 | 5 | 775 | |||
GenBank mRNA | AK057345.1 | - | 5 | 533 | ||
AK290346.1 | - | 5 | 517 | |||
AK292871.1 | - | 5 | 522 | |||
BC036767.1 | - | 5 | 482 | 5744127 |