Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152542.4 | NP_689755.3 | 7 | 1709 | ||
XM_006714111.3 | XP_006714174.1 | 7 | 1770 | |||
XM_017007803.1 | XP_016863292.1 | 7 | 1357 | |||
XM_017007804.1 | XP_016863293.1 | 8 | 1897 | |||
XM_017007805.1 | XP_016863294.1 | 9 | 1897 | |||
XM_017007806.1 | XP_016863295.1 | 7 | 1230 | |||
XM_017007807.1 | XP_016863296.1 | 6 | 948 | |||
GenBank mRNA | AK054678.1 | - | 7 | 1437 | ||
AL834271.1 | - | 7 | 1428 | |||
AY157615.1 | - | 7 | 1437 | |||
AY435431.1 | - | 7 | 1614 | |||
BC037552.1 | - | 7 | 1473 | 5227317 |