Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287501.1 | NP_001274430.1 | 20 | 2305 | ||
NM_152558.4 | NP_689771.3 | 22 | 2399 | |||
XM_017011902.1 | XP_016867391.1 | 22 | 2461 | |||
XM_017011903.1 | XP_016867392.1 | 22 | 2339 | |||
GenBank mRNA | AB028946.2 | - | 20 | 2102 | ||
AL137525.1 | - | 1 | 120 | |||
BC043150.1 | - | 20 | 2225 | 5297871 | ||
BC071858.1 | - | 22 | 2373 | 5478116 | ||
DA246430.1 | - | 5 | 577 |