Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110199.2 | NP_001103669.1 | 15 | 2440 | ||
NM_001291831.1 | NP_001278760.1 | 16 | 2519 | |||
GenBank mRNA | AK092590.1 | - | 3 | 858 | ||
AK094397.1 | - | 1 | 1051 | |||
BC063788.1 | - | 1 | 779 | |||
BC096755.1 | - | 4 | 969 |