Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALYL SIRNA Assays ›
Gene Symbol:
RALYL
Gene Name:
RALY RNA binding protein-like
Gene Aliases:
HNRPCL3
Chromosome Location:
Chr.8: 84182629 - 84921844 on Build GRCh38
UniGene ID:
Hs.121663
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128318
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALYL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100391.1
NP_001093861.1
2, 3
431
NM_001100392.1
NP_001093862.1
3, 4
674
NM_001100393.1
NP_001093863.1
2, 3
642
NM_001287243.1
NP_001274172.1
2, 3
1355
NM_173848.5
NP_776247.3
2, 3
1355
XM_011517462.2
XP_011515764.1
3, 4
1459
XM_017013058.1
XP_016868547.1
2, 3
630
XM_017013059.1
XP_016868548.1
4, 5
725
XM_017013060.1
XP_016868549.1
2, 3
2875
XM_017013061.1
XP_016868550.1
2, 3
2027
XM_017013062.1
XP_016868551.1
4, 5
834
XM_017013063.1
XP_016868552.1
2, 3
597
XM_017013064.1
XP_016868553.1
3, 4
948
XM_017013065.1
XP_016868554.1
2, 3
496
XM_017013066.1
XP_016868555.1
2, 3
642
XM_017013067.1
XP_016868556.1
3, 4
1533
XM_017013068.1
XP_016868557.1
4, 5
725
XM_017013069.1
XP_016868558.1
4, 5
781
XM_017013070.1
XP_016868559.1
3, 4
625
XM_017013071.1
XP_016868560.1
2, 3
496
XM_017013072.1
XP_016868561.1
2, 3
583
XM_017013073.1
XP_016868562.1
2, 3
583
XM_017013074.1
XP_016868563.1
2, 3
496
XM_017013075.1
XP_016868564.1
2, 3
581
XM_017013076.1
XP_016868565.1
2, 3
376
XM_017013077.1
XP_016868566.1
3, 4
1626
XM_017013078.1
XP_016868567.1
2, 3
642
XM_017013079.1
XP_016868568.1
3, 4
625
XM_017013080.1
XP_016868569.1
2, 3
376
XM_017013081.1
XP_016868570.1
2, 3
376
XM_017013082.1
XP_016868571.1
2, 3
642
GenBank mRNA
AF495529.1
-
2, 3
1355
AF495530.1
-
2, 3
572
AK094543.1
-
2, 3
586
AK095026.1
-
2, 3
1171
AK095560.1
-
3, 4
625
BC031090.1
-
2, 3
420
5311935
DN989439.1
-
2, 3
436
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0