Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177965.3 | NP_808880.1 | 6 | 903 | ||
XM_005250799.3 | XP_005250856.2 | 5 | 2175 | |||
GenBank mRNA | AK055162.1 | - | 6 | 878 | ||
BC036855.2 | - | 6 | 868 | |||
GQ240139.1 | - | 6 | 877 |