Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039803.2 | NP_001034892.1 | 6 | 966 | ||
NM_001170639.1 | NP_001164110.1 | 5 | 903 | |||
NM_012119.4 | NP_036251.2 | 5 | 903 | |||
NM_178432.3 | NP_848519.1 | 5 | 942 | |||
XM_017014561.1 | XP_016870050.1 | 6 | 911 | |||
XM_017014562.1 | XP_016870051.1 | 6 | 911 | |||
XM_017014564.1 | XP_016870053.1 | 5 | 833 | |||
GenBank mRNA | AF035013.1 | - | 5 | 806 | ||
AF113130.1 | - | 4 | 1670 | |||
AK075325.1 | - | 5 | 695 | |||
AK226135.1 | - | 2 | 2098 | |||
AK294193.1 | - | 5 | 864 | |||
BC002655.2 | - | 5 | 628 | 3605918 | ||
BX647489.1 | - | 4 | 1688 | |||
KU178572.1 | - | 5 | 569 | |||
KU178573.1 | - | 5 | 569 | |||
KU178574.1 | - | 6 | 632 | |||
KU178575.1 | - | 5 | 569 |