Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178819.3 | NP_848934.1 | 7 | 1698 | ||
XM_011544392.2 | XP_011542694.1 | 8 | 1821 | |||
XM_011544393.1 | XP_011542695.1 | 7 | 1710 | |||
XM_011544394.1 | XP_011542696.1 | 7 | 1747 | |||
XM_017013042.1 | XP_016868531.1 | 8 | 1848 | |||
XM_017013043.1 | XP_016868532.1 | 8 | 1719 | |||
GenBank mRNA | AF317516.1 | - | 6 | 924 | ||
AF406612.1 | - | 7 | 1686 | |||
AL834255.1 | - | 3 | 183 | |||
AY358670.1 | - | 6 | 835 | |||
AY513610.1 | - | 6 | 918 | |||
BC051377.1 | - | 6 | 878 | 6649895 | ||
BC061884.1 | - | 6 | 878 | 6577569 |