Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143944.1 | NP_001137416.1 | 8 | 1212 | ||
NM_181336.3 | NP_851853.1 | 9 | 1793 | |||
GenBank mRNA | AK024858.1 | - | 3 | 619 | ||
AK026077.1 | - | 1 | 59 | |||
AK074959.1 | - | 9 | 1846 | |||
AK127084.1 | - | 5 | 3734 | |||
AK301083.1 | - | 8 | 1206 | |||
AK309888.1 | - | 5 | 970 | |||
BC051803.1 | - | 9 | 1783 | 5240212 |