Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143944.1 | NP_001137416.1 | 8 | 1466 | ||
NM_181336.3 | NP_851853.1 | 9 | 2047 | |||
GenBank mRNA | AK024858.1 | - | 3 | 873 | ||
AK026077.1 | - | 1 | 313 | |||
AK074959.1 | - | 9 | 2100 | |||
AK127084.1 | - | 5 | 3988 | |||
BC051803.1 | - | 9 | 2037 | 5240212 |