Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003246.3 | NP_003237.2 | 3 | 593 | ||
XM_011521971.1 | XP_011520273.1 | 3 | 589 | |||
GenBank mRNA | AB209912.1 | - | 3 | 579 | ||
AK291639.1 | - | 3 | 593 | |||
AK304754.1 | - | 3 | 339 | |||
BC028145.1 | - | 2 | 448 | |||
BC136469.1 | - | 3 | 567 | 9020458 | ||
BC136470.1 | - | 3 | 567 | 9020459 | ||
M14326.1 | - | 3 | 534 | |||
M25631.1 | - | 3 | 513 | |||
X04665.1 | - | 3 | 489 | |||
X14787.1 | - | 3 | 525 |