Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105543.1 | NP_001099013.1 | 2 | 348 | ||
NM_020911.1 | NP_065962.1 | 2 | 348 | |||
NM_181775.3 | NP_861440.2 | 3 | 496 | |||
XM_005250686.4 | XP_005250743.1 | 2 | 746 | |||
XM_006716171.3 | XP_006716234.1 | 3 | 839 | |||
XM_011516676.2 | XP_011514978.1 | 2 | 254 | |||
XM_017012779.1 | XP_016868268.1 | 2 | 254 | |||
GenBank mRNA | AK123428.1 | - | 2 | 254 | ||
BC028744.2 | - | 3 | 496 |