Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEPACAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039372.2 | NP_001034461.1 | 2, 3 | 503 | ||
NM_001288804.1 | NP_001275733.1 | 3, 4 | 572 | |||
NM_001288810.1 | NP_001275739.1 | 1, 2 | 495 | |||
NM_198151.2 | NP_937794.1 | 1, 2 | 495 | |||
XM_011516000.2 | XP_011514302.1 | 2, 3 | 436 | |||
XM_011516001.1 | XP_011514303.1 | 2, 3 | 436 | |||
GenBank mRNA | AK096002.1 | - | 2, 3 | 456 | ||
AK298361.1 | - | 3, 4 | 522 | |||
AY358345.1 | - | 1, 2 | 457 | |||
BC136300.1 | - | 1, 2 | 425 | 9020287 | ||
BC136301.1 | - | 1, 2 | 425 | 9020288 | ||
BC139906.1 | - | 1, 2 | 482 | 40147445 | ||
DA426282.1 | - | 2, 3 | 503 |