Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288564.1 | NP_001275493.1 | 6 | 871 | ||
NM_001288565.1 | NP_001275494.1 | 5 | 1374 | |||
NM_001288566.1 | NP_001275495.1 | 6 | 972 | |||
NM_001288567.1 | NP_001275496.1 | 6 | 1186 | |||
NM_001288568.1 | NP_001275497.1 | 6 | 1024 | |||
NM_001288569.1 | NP_001275498.1 | 6 | 885 | |||
NM_001288570.1 | NP_001275499.1 | 6 | 898 | |||
NM_001288571.1 | NP_001275500.1 | 6 | 964 | |||
NM_016456.4 | NP_057540.1 | 6 | 889 | |||
NR_109920.1 | 7 | 1116 | ||||
NR_109921.1 | 5 | 1343 | ||||
XM_011509385.1 | XP_011507687.1 | 6 | 948 | |||
XM_011509386.2 | XP_011507688.1 | 6 | 923 | |||
XM_011509387.2 | XP_011507689.1 | 6 | 811 | |||
GenBank mRNA | AF151020.1 | - | 7 | 861 | ||
AK054883.1 | - | 6 | 844 | |||
AK075335.1 | - | 6 | 799 | |||
AK091726.1 | - | 5 | 1259 | |||
AK094588.1 | - | 2 | 2192 | |||
AK303843.1 | - | 6 | 910 | |||
AL136600.1 | - | 6 | 828 | |||
AY359012.1 | - | 6 | 863 | |||
BC001106.2 | - | 6 | 813 | 3506442 |