Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270695.1 | NP_001257624.1 | 6 | 1410 | ||
NM_001270696.1 | NP_001257625.1 | 6 | 1410 | |||
NM_001270697.1 | NP_001257626.1 | 5 | 1269 | |||
NM_001270698.1 | NP_001257627.1 | 5 | 1269 | |||
NM_001270699.1 | NP_001257628.1 | 3 | 1036 | |||
NM_018287.6 | NP_060757.4 | 6 | 1410 | |||
XM_005252644.1 | XP_005252701.1 | 5 | 1269 | |||
GenBank mRNA | AK000744.1 | - | 3 | 478 | ||
AK091182.1 | - | 6 | 1355 | |||
AK127611.1 | - | 5 | 1311 | |||
AK315066.1 | - | 5 | 1162 | |||
AL834250.2 | - | 3 | 960 | |||
AY033594.1 | - | 6 | 1377 | |||
AY033595.1 | - | 6 | 1377 | |||
BC094719.1 | - | 6 | 1334 | 30528830 | ||
BC115362.2 | - | 5 | 1166 | 40035586 | ||
BC115363.2 | - | 5 | 1166 | 40035587 | ||
CR936681.1 | - | 7 | 1311 |