Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033198.3 | NP_149975.1 | 7 | 1150 | ||
GenBank mRNA | AB057723.1 | - | 7 | 777 | ||
AF161427.1 | - | 1 | 12 | |||
AK297046.1 | - | 6 | 669 | |||
AK314583.1 | - | 7 | 813 | |||
AY359112.1 | - | 6 | 1045 | |||
BC007301.1 | - | 5 | 534 | |||
BC069228.1 | - | 7 | 800 | 3944034 |