Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052876.3 | NP_443108.1 | 4 | 1307 | ||
XM_005259721.3 | XP_005259778.1 | 5 | 1366 | |||
GenBank mRNA | AF395817.1 | - | 4 | 1258 | ||
AK226064.1 | - | 2 | 114 | |||
BC055396.1 | - | 5 | 1241 | 6142288 |