Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281503.1 | NP_001268432.1 | 2 | 3535 | ||
NM_052910.2 | NP_443142.1 | 1 | 3978 | |||
GenBank mRNA | AB067497.1 | - | 1 | 3977 | ||
BC051738.1 | - | 1 | 3879 | 4816570 |