Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052924.2 | NP_443156.2 | 14 | 1794 | ||
XM_005250772.3 | XP_005250829.1 | 15 | 1980 | |||
XM_005250773.3 | XP_005250830.1 | 15 | 1977 | |||
XM_006716502.3 | XP_006716565.1 | 15 | 1995 | |||
XM_011516817.2 | XP_011515119.1 | 15 | 1998 | |||
XM_011516818.1 | XP_011515120.1 | 15 | 2655 | |||
XM_011516819.2 | XP_011515121.1 | 14 | 1839 | |||
XM_011516820.2 | XP_011515122.1 | 14 | 2182 | |||
XM_011516821.2 | XP_011515123.1 | 14 | 2186 | |||
XM_011516822.2 | XP_011515124.1 | 14 | 2331 | |||
XM_011516823.2 | XP_011515125.1 | 14 | 1922 | |||
XM_017013013.1 | XP_016868502.1 | 14 | 1870 | |||
XM_017013014.1 | XP_016868503.1 | 14 | 1919 | |||
XM_017013015.1 | XP_016868504.1 | 14 | 2312 | |||
XM_017013016.1 | XP_016868505.1 | 14 | 1903 | |||
GenBank mRNA | AB067516.1 | - | 13 | 1728 | ||
AY082588.1 | - | 13 | 1736 | |||
BC025767.1 | - | 14 | 1762 | 5211354 |