Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052941.4 | NP_443173.2 | 11 | 2834 | ||
GenBank mRNA | AK131094.1 | - | 11 | 2747 | ||
AL832576.2 | - | 11 | 2765 | |||
BC070055.1 | - | 11 | 2771 | 30345441 |