Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP6K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142883.1 | NP_001136355.1 | 7 | 2472 | ||
NM_054111.4 | NP_473452.2 | 6 | 2343 | |||
XM_005248842.3 | XP_005248899.1 | 7 | 2252 | |||
XM_005248843.3 | XP_005248900.1 | 5 | 2363 | |||
GenBank mRNA | AF393812.1 | - | 5 | 2038 | ||
AK056586.2 | - | 6 | 2262 | |||
AK223572.1 | - | 6 | 2266 | |||
BC089389.1 | - | 6 | 2343 | 30530548 |