Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078483.3 | NP_510968.2 | 11 | 2962 | ||
XM_005268386.1 | XP_005268443.1 | 11 | 2932 | |||
XM_006714759.3 | XP_006714822.1 | 11 | 3011 | |||
XM_011537580.2 | XP_011535882.1 | 11 | 3652 | |||
XM_011537581.1 | XP_011535883.1 | 12 | 3080 | |||
XM_011537583.2 | XP_011535885.1 | 12 | 7517 | |||
XM_011537584.2 | XP_011535886.1 | 11 | 3171 | |||
XM_011537585.1 | XP_011535887.1 | 12 | 3027 | |||
XM_011537586.2 | XP_011535888.1 | 11 | 3775 | |||
XM_011537587.2 | XP_011535889.1 | 11 | 3031 | |||
XM_011537588.2 | XP_011535890.1 | 11 | 2875 | |||
XM_011537589.2 | XP_011535891.1 | 11 | 3783 | |||
XM_011537590.1 | XP_011535892.1 | 11 | 2790 | |||
XM_011537591.1 | XP_011535893.1 | 11 | 2765 | |||
XM_011537592.2 | XP_011535894.1 | 11 | 2769 | |||
XM_011537593.2 | XP_011535895.1 | 12 | 3025 | |||
XM_011537594.1 | XP_011535896.1 | 13 | 3139 | |||
XM_017009216.1 | XP_016864705.1 | 11 | 2858 | |||
GenBank mRNA | AF516142.1 | - | 10 | 2754 | ||
AK025102.1 | - | 1 | 534 | |||
AL833703.1 | - | 1 | 227 | |||
BC136437.1 | - | 11 | 2833 | 9020426 | ||
BC136438.1 | - | 11 | 2833 | 9020427 | ||
BX537963.1 | - | 11 | 2959 |