Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TJAP1 SIRNA Assays ›
Gene Symbol:
TJAP1
Gene Name:
tight junction associated protein 1
Gene Aliases:
PILT, TJP4
Chromosome Location:
Chr.6: 43477523 - 43506557 on Build GRCh38
UniGene ID:
Hs.520145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128982
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TJAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146016.1
NP_001139488.1
11
2462
NM_001146017.1
NP_001139489.1
10
2427
NM_001146018.1
NP_001139490.1
11
2489
NM_001146019.1
NP_001139491.1
10
2432
NM_001146020.1
NP_001139492.1
9
2283
NM_080604.2
NP_542171.2
10
2484
NR_027394.1
10
2349
XM_006715249.1
XP_006715312.1
11
2440
XM_006715250.3
XP_006715313.1
12
2459
XM_006715251.2
XP_006715314.1
11
2467
XM_006715252.2
XP_006715315.1
12
2527
XM_006715254.3
XP_006715317.1
11
2365
XM_006715256.1
XP_006715319.1
11
2362
XM_006715257.2
XP_006715320.1
11
2416
XM_006715259.3
XP_006715322.1
10
2266
XM_006715260.1
XP_006715323.1
11
2383
XM_006715261.1
XP_006715324.1
10
2287
XM_006715262.2
XP_006715325.1
11
2612
XM_006715263.2
XP_006715326.1
10
2322
XM_006715265.1
XP_006715328.1
10
2332
XM_006715266.1
XP_006715329.1
9
2240
XM_006715267.2
XP_006715330.1
11
2451
XM_006715268.2
XP_006715331.1
11
2448
XM_006715269.2
XP_006715332.1
11
2503
XM_006715270.1
XP_006715333.1
10
2421
XM_011514995.1
XP_011513297.1
12
2502
XM_011514996.1
XP_011513298.1
11
2422
XM_017011488.1
XP_016866977.1
10
2340
XM_017011489.1
XP_016866978.1
11
2637
XM_017011490.1
XP_016866979.1
10
2410
XM_017011491.1
XP_016866980.1
11
2446
XM_017011492.1
XP_016866981.1
12
2575
XM_017011493.1
XP_016866982.1
12
2573
GenBank mRNA
AJ420477.1
-
2
1583
AK024269.1
-
10
2434
AK074241.1
-
5
1878
AK094907.1
-
3
2290
AK123398.1
-
7
4992
AK127338.1
-
9
3545
AL117398.1
-
4
2916
BC046239.1
-
11
2382
4906152
BC064401.1
-
10
2363
5539930
Back To Top
More Information
Panther Classification:
Molecular Function -
tight junction
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0