Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080751.2 | NP_542789.2 | 11, 12 | 1436 | ||
XM_005260660.3 | XP_005260717.1 | 9, 10 | 1630 | |||
GenBank mRNA | AF417580.2 | - | 11, 12 | 1436 | ||
AK094789.1 | - | 1, 2 | 144 | |||
AK294013.1 | - | 9, 10 | 1182 | |||
AK316261.1 | - | 9, 10 | 1152 |