Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080868.2 | NP_543144.1 | 2 | 560 | ||
XM_006710345.3 | XP_006710408.1 | 3 | 392 | |||
XM_006710346.3 | XP_006710409.1 | 3 | 319 | |||
GenBank mRNA | AF403035.1 | - | 2 | 420 | ||
AF429968.1 | - | 2 | 537 | |||
AK098606.1 | - | 2 | 560 | |||
BC036573.2 | - | 2 | 538 | 5297837 |