Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322818.1 | NP_001309747.1 | 10 | 1447 | ||
NM_080872.3 | NP_543148.2 | 10 | 1734 | |||
XM_011544400.2 | XP_011542702.1 | 9 | 1675 | |||
XM_011544401.2 | XP_011542703.1 | 10 | 1669 | |||
XM_011544402.2 | XP_011542704.1 | 9 | 1540 | |||
XM_011544403.2 | XP_011542705.1 | 8 | 1507 | |||
XM_011544404.2 | XP_011542706.1 | 9 | 1501 | |||
XM_017013044.1 | XP_016868533.1 | 11 | 1781 | |||
XM_017013045.1 | XP_016868534.1 | 11 | 1751 | |||
XM_017013046.1 | XP_016868535.1 | 10 | 1748 | |||
XM_017013047.1 | XP_016868536.1 | 10 | 1612 | |||
XM_017013048.1 | XP_016868537.1 | 10 | 2894 | |||
XM_017013049.1 | XP_016868538.1 | 10 | 2845 | |||
XM_017013050.1 | XP_016868539.1 | 11 | 2301 | |||
GenBank mRNA | AB055056.1 | - | 10 | 1446 | ||
AK122610.1 | - | 2 | 245 | |||
AK122615.1 | - | 5 | 768 | |||
AY358147.1 | - | 10 | 1734 |