Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080874.3 | NP_543150.1 | 7 | 1580 | ||
XM_005262759.1 | XP_005262816.1 | 9 | 2009 | |||
XM_011531616.2 | XP_011529918.1 | 7 | 1688 | |||
XM_011531617.2 | XP_011529919.1 | 7 | 2271 | |||
GenBank mRNA | AK096441.1 | - | 7 | 1482 | ||
BX647857.1 | - | 7 | 1579 |