Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277961.1 | NP_001264890.1 | 12 | 3881 | ||
NM_001277962.1 | NP_001264891.1 | 13 | 3600 | |||
NM_003156.3 | NP_003147.2 | 12 | 3563 | |||
GenBank mRNA | BC021300.2 | - | 12 | 3555 | 4899542 | |
CX873095.1 | - | 1 | 394 | |||
HQ317451.1 | - | 12 | 3880 | |||
U52426.1 | - | 12 | 3558 |