Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBED6CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138434.2 | NP_612443.1 | 1 | 737 | ||
GenBank mRNA | AL832660.1 | - | 1 | 796 | ||
BC011406.1 | - | 1 | 274 | 3855620 |