Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322250.1 | NP_001309179.1 | 2, 3 | 172 | ||
NM_001322251.1 | NP_001309180.1 | 2, 3 | 276 | |||
NM_138962.3 | NP_620412.1 | 2, 3 | 276 | |||
NM_170721.1 | NP_733839.1 | 1, 2 | 126 | |||
XM_005257014.3 | XP_005257071.1 | 2, 3 | 201 | |||
XM_005257015.3 | XP_005257072.1 | 2, 3 | 201 | |||
XM_011524286.1 | XP_011522588.1 | 2, 3 | 114 | |||
XM_017024147.1 | XP_016879636.1 | 2, 3 | 143 | |||
XM_017024148.1 | XP_016879637.1 | 2, 3 | 201 | |||
GenBank mRNA | AK093888.1 | - | 1, 2 | 126 | ||
AK295060.1 | - | 2, 3 | 173 | |||
BC001526.2 | - | 2, 3 | 276 | 3505639 |