Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308292.1 | NP_001295221.1 | 3 | 830 | ||
NM_144727.2 | NP_653328.1 | 4 | 819 | |||
NR_131779.1 | 4 | 1171 | ||||
GenBank mRNA | AF445455.1 | - | 4 | 662 | ||
BC043605.1 | - | 4 | 1171 | |||
BC100878.2 | - | 4 | 657 | 40003204 | ||
BC100879.3 | - | 3 | 668 | 40003207 | ||
BC100880.2 | - | 4 | 657 | 40003208 | ||
BC100881.2 | - | 4 | 657 | 40003209 |