Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCNY SIRNA Assays ›
Gene Symbol:
CCNY
Gene Name:
cyclin Y
Gene Aliases:
C10orf9, CBCP1, CCNX, CFP1
Chromosome Location:
Chr.10: 35246981 - 35572670 on Build GRCh38
UniGene ID:
Hs.14745
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
129280
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCNY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282852.1
NP_001269781.1
1, 2
341
NM_001282853.1
NP_001269782.1
3, 4
479
NM_001282854.1
NP_001269783.1
2, 3
416
NM_145012.5
NP_659449.3
2, 3
416
NM_181698.3
NP_859049.2
4, 5
440
XM_006717392.3
XP_006717455.1
2, 3
185
XM_011519357.2
XP_011517659.1
2, 3
411
XM_011519358.1
XP_011517660.1
5, 6
507
XM_011519360.2
XP_011517662.1
6, 7
609
XM_011519361.1
XP_011517663.1
6, 7
491
XM_011519362.1
XP_011517664.1
5, 6
471
XM_011519363.1
XP_011517665.1
5, 6
484
XM_011519364.1
XP_011517666.1
7, 8
605
XM_011519365.2
XP_011517667.1
6, 7
524
XM_011519366.1
XP_011517668.1
5, 6
454
XM_011519367.1
XP_011517669.1
3, 4
284
XM_011519368.2
XP_011517670.1
2, 3
179
XM_011519369.2
XP_011517671.1
2, 3
770
XM_017015839.1
XP_016871328.1
2, 3
102
XM_017015840.1
XP_016871329.1
2, 3
104
XM_017015841.1
XP_016871330.1
2, 3
102
GenBank mRNA
AF413522.1
-
2, 3
416
AF429969.1
-
4, 5
396
AF465729.1
-
2, 3
237
AK057280.1
-
4, 5
396
AK314666.1
-
3, 4
455
AL834355.1
-
2, 3
274
AY504868.1
-
2, 3
416
BC069224.1
-
2, 3
242
BC094815.1
-
2, 3
268
6340750
BC104773.1
-
3, 4
225
8143776
BC104801.1
-
3, 4
225
8143804
BC143450.1
-
2, 3
118
9051958
BC143455.1
-
3, 4
225
9051963
DB037034.1
-
5, 6
471
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026484, AK017493, BC035524, AK076556, BC023321, and others.
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
Wnt signaling pathway
positive regulation of cyclin-dependent protein serine/threonine kinase activity
cell division
regulation of canonical Wnt signaling pathway
protein binding
cyclin-dependent protein serine/threonine kinase regulator activity
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0